siRNA Details
| Gene Symbol: GTF2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278940.1 | NP_001265869.1 | 10 | 3574 | ||
| NM_015859.3 | NP_056943.1 | 9 | 3507 | |||
| NM_201595.2 | NP_963889.1 | 9 | 3127 | |||
| GenBank mRNA | AK025178.1 | - | 1 | 190 | ||
| BC041410.2 | - | 1 | 905 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC037828.1 | - | 1 | 1722 | Chr. 14 - 81176140 - 81180000 [-] on Build GRCh38 | |

GTF2A1
Genomic Map