siRNA Details
| Gene Symbol: GTF2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278940.1 | NP_001265869.1 | 10 | 2442 | ||
| NM_015859.3 | NP_056943.1 | 9 | 2375 | |||
| NM_201595.2 | NP_963889.1 | 9 | 1995 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GTF2A1| Gene Symbol: GTF2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278940.1 | NP_001265869.1 | 10 | 2442 | ||
| NM_015859.3 | NP_056943.1 | 9 | 2375 | |||
| NM_201595.2 | NP_963889.1 | 9 | 1995 | |||