siRNA Details
| Gene Symbol: THEGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011534357.1 | XP_011532659.1 | 11 | 1695 | ||
| GenBank mRNA | AL833514.1 | - | 2 | 3111 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

THEGL| Gene Symbol: THEGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011534357.1 | XP_011532659.1 | 11 | 1695 | ||
| GenBank mRNA | AL833514.1 | - | 2 | 3111 | ||