siRNA Details
| Gene Symbol: LINC00029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL117533.1 | - | 3 | 1167 | ||
| BC134343.1 | - | 2 | 335 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX101555.1 | - | 4 | 463 | Chr. 20 - 63034349 - 63036139 [-] on Build GRCh38 | |

LINC00029
Genomic Map