siRNA Details
Gene Symbol: LINC00029 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL117533.1 | - | 3 | 1328 | ||
BC134343.1 | - | 2 | 496 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX101555.1 | - | 4 | 624 | Chr. 20 - 63034349 - 63036139 [-] on Build GRCh38 |
