siRNA Details
| Gene Symbol: CEP41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257158.1 | NP_001244087.1 | 10 | 4736 | ||
| NM_001257159.1 | NP_001244088.1 | 9 | 4688 | |||
| NM_018718.2 | NP_061188.1 | 11 | 4952 | |||
| NR_046443.1 | 11 | 4876 | ||||
| GenBank mRNA | BC047500.1 | - | 1 | 2454 | ||

CEP41
Genomic Map