siRNA Details
| Gene Symbol: CEP41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257158.1 | NP_001244087.1 | 10 | 4213 | ||
| NM_001257159.1 | NP_001244088.1 | 9 | 4165 | |||
| NM_018718.2 | NP_061188.1 | 11 | 4429 | |||
| NR_046443.1 | 11 | 4353 | ||||
| XM_011516709.2 | XP_011515011.1 | 12 | 4390 | |||
| XM_011516710.2 | XP_011515012.1 | 10 | 4205 | |||
| GenBank mRNA | BC047500.1 | - | 1 | 1930 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CK001131.1 | - | 1 | 363 | Chr. 7 - 130395535 - 130396189 [-] on Build GRCh38 | |

CEP41
Genomic Map