siRNA Details
| Gene Symbol: LOC101929333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127909.1 | - | 2 | 441 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA940586.1 | - | 2 | 441 | Chr. 15 - 74480863 - 74490286 [-] on Build GRCh38 | |

LOC101929333
Genomic Map