siRNA Details
| Gene Symbol: TTC21B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK123487.1 | - | 5 | 2336 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTC21B-AS1| Gene Symbol: TTC21B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK123487.1 | - | 5 | 2336 | ||