siRNA Details
| Gene Symbol: SCARA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017013537.1 | XP_016869026.1 | 7 | 2276 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK130667.1 | - | 1 | 2407 | Chr. 8 - 27717072 - 27719537 [+] on Build GRCh38 | |

SCARA3
Genomic Map