siRNA Details
| Gene Symbol: DCUN1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308101.1 | NP_001295030.1 | 7 | 7482 | ||
| NM_020640.3 | NP_065691.2 | 7 | 7464 | |||
| XM_011512912.1 | XP_011511214.1 | 7 | 2799 | |||
| XM_011512914.1 | XP_011511216.1 | 7 | 2838 | |||
| XM_011512915.2 | XP_011511217.1 | 8 | 3360 | |||
| XM_011512916.2 | XP_011511218.1 | 7 | 3288 | |||
| XM_017006633.1 | XP_016862122.1 | 7 | 2610 | |||
| XM_017006634.1 | XP_016862123.1 | 7 | 2804 | |||
| GenBank mRNA | AK095773.1 | - | 1 | 2168 | ||

DCUN1D1
Genomic Map