siRNA Details
| Gene Symbol: DCUN1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308101.1 | NP_001295030.1 | 7 | 5394 | ||
| NM_020640.3 | NP_065691.2 | 7 | 5376 | |||
| GenBank mRNA | AK095773.1 | - | 1 | 80 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DCUN1D1| Gene Symbol: DCUN1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308101.1 | NP_001295030.1 | 7 | 5394 | ||
| NM_020640.3 | NP_065691.2 | 7 | 5376 | |||
| GenBank mRNA | AK095773.1 | - | 1 | 80 | ||