siRNA Details
Gene Symbol: ZNF800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250181.3 | XP_005250238.1 | 6 | 11133 | ||
GenBank mRNA | BX537723.1 | - | 1 | 1287 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250181.3 | XP_005250238.1 | 6 | 11133 | ||
GenBank mRNA | BX537723.1 | - | 1 | 1287 |