siRNA Details
| Gene Symbol: ZNF800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005250181.3 | XP_005250238.1 | 6 | 13607 | ||
| GenBank mRNA | BX537723.1 | - | 1 | 3761 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF136125.1 | - | 1 | 197 | Chr. 7 - 127360574 - 127361188 [-] on Build GRCh38 | |

ZNF800
Genomic Map