siRNA Details
| Gene Symbol: ZNF800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005250181.3 | XP_005250238.1 | 6 | 14375 | ||
| GenBank mRNA | BX537723.1 | - | 1 | 4529 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF800| Gene Symbol: ZNF800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005250181.3 | XP_005250238.1 | 6 | 14375 | ||
| GenBank mRNA | BX537723.1 | - | 1 | 4529 | ||