siRNA Details
| Gene Symbol: LOC101927914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110157.1 | 1 | 1471 | |||
| GenBank mRNA | BC039304.1 | - | 1 | 1474 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000444158.1 | - | 1 | 1471 | Chr. 7 - 157466231 - 157499716 [+] on Build GRCh38 | |

LOC101927914
Genomic Map