siRNA Details
| Gene Symbol: LOC101927914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110157.1 | 1 | 233 | |||
| GenBank mRNA | BC039304.1 | - | 1 | 236 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927914| Gene Symbol: LOC101927914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110157.1 | 1 | 233 | |||
| GenBank mRNA | BC039304.1 | - | 1 | 236 | ||