siRNA Details
| Gene Symbol: LOC286191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL833212.1 | - | 1 | 827 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE067934.1 | - | 1 | 152 | Chr. 8 - 73061997 - 73062527 [-] on Build GRCh38 | |

LOC286191
Genomic Map