siRNA Details
Gene Symbol: LINC00560 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC043278.2 | - | 2 | 1556 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000564593.1 | - | 2 | 1556 | Chr. 13 - 88626853 - 88630397 [+] on Build GRCh38 |
