siRNA Details
| Gene Symbol: LINC00560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC043278.2 | - | 1 | 304 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00560| Gene Symbol: LINC00560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC043278.2 | - | 1 | 304 | ||