siRNA Details
| Gene Symbol: LOC100130744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK129556.1 | - | 1 | 1020 | ||
| Gene Symbol: OTULIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011514151.2 | XP_011512453.1 | 8 | 8115 | ||

LOC100130744
Genomic Map