siRNA Details
| Gene Symbol: ZNF497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | DA358776.1 | - | 1 | 540 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK125350.1 | - | 1 | 442 | Chr. 19 - 58360801 - 58362751 [-] on Build GRCh38 | |

ZNF497
Genomic Map