siRNA Details
| Gene Symbol: CLEC19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256720.1 | NP_001243649.1 | 5 | 1572 | ||
| GenBank mRNA | BX640722.1 | - | 3 | 1459 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLEC19A| Gene Symbol: CLEC19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256720.1 | NP_001243649.1 | 5 | 1572 | ||
| GenBank mRNA | BX640722.1 | - | 3 | 1459 | ||