siRNA Details
| Gene Symbol: MIATNB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK026502.1 | - | 2 | 593 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB126006.1 | - | 2 | 546 | Chr. 22 - 26672916 - 26717736 [+] on Build GRCh38 | |

MIATNB
Genomic Map