siRNA Details
| Gene Symbol: LOC101927845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125427.1 | 1 | 413 | |||
| GenBank mRNA | AK057402.1 | - | 1 | 413 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927845| Gene Symbol: LOC101927845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125427.1 | 1 | 413 | |||
| GenBank mRNA | AK057402.1 | - | 1 | 413 | ||