siRNA Details
| Gene Symbol: CPPED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX092044.1 | - | 1 | 186 | ||
| Gene Symbol: LINC00868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX089989.1 | - | 2 | 254 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK056117.1 | - | 1 | 21 | Chr. 17 - 65805972 - 65808621 [-] on Build GRCh38 | |

CPPED1
Genomic Map