siRNA Details
| Gene Symbol: SMCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ230805.1 | - | 1 | 479 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMCR6| Gene Symbol: SMCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ230805.1 | - | 1 | 479 | ||