siRNA Details
| Gene Symbol: TEKT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005263876.2 | XP_005263933.1 | 1 | 401 | ||
| XM_011510670.1 | XP_011508972.1 | 1 | 401 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR617898.1 | - | 1 | 682 | Chr. 2 - 94867486 - 94868761 [+] on Build GRCh38 | |

TEKT4
Genomic Map