siRNA Details
| Gene Symbol: PPIE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006112.3 | NP_006103.1 | 10 | 4326 | ||
| NR_036543.1 | 10 | 4318 | ||||
| NR_036544.1 | 10 | 4279 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ187153.1 | - | 1 | 549 | Chr. 1 - 39756172 - 39756771 [+] on Build GRCh38 | |

PPIE
Genomic Map