siRNA Details
| Gene Symbol: PPIE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006112.3 | NP_006103.1 | 10 | 4257 | ||
| NR_036543.1 | 10 | 4249 | ||||
| NR_036544.1 | 10 | 4210 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPIE| Gene Symbol: PPIE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006112.3 | NP_006103.1 | 10 | 4257 | ||
| NR_036543.1 | 10 | 4249 | ||||
| NR_036544.1 | 10 | 4210 | ||||