siRNA Details
| Gene Symbol: LVCAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131958.1 | 2 | 2085 | |||
| NR_131959.1 | 2 | 1978 | ||||
| GenBank mRNA | BC042992.1 | - | 2 | 2090 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LVCAT1| Gene Symbol: LVCAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131958.1 | 2 | 2085 | |||
| NR_131959.1 | 2 | 1978 | ||||
| GenBank mRNA | BC042992.1 | - | 2 | 2090 | ||