siRNA Details
| Gene Symbol: GPR107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287346.1 | NP_001274275.1 | 17 | 2787 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK023462.1 | - | 1 | 125 | Chr. 9 - 130129639 - 130132793 [+] on Build GRCh38 | |
More Information
Set Membership: |
|

GPR107
Genomic Map