siRNA Details
| Gene Symbol: DISP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011522125.2 | XP_011520427.1 | 1 | 640 | ||
| XM_017022687.1 | XP_016878176.1 | 1 | 640 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK130410.1 | - | 1 | 1299 | Chr. 15 - 40353699 - 40355173 [+] on Build GRCh38 | |

DISP2
Genomic Map