siRNA Details
Gene Symbol: ESRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU934976.1 | - | 1 | 79 |
Gene Symbol: NFATC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004555.3 | NP_004546.1 | 11 | 5673 | ||
NM_173163.2 | NP_775186.1 | 11 | 5658 | |||
NM_173165.2 | NP_775188.1 | 10 | 5569 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
