siRNA Details
| Gene Symbol: NFATC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004555.3 | NP_004546.1 | 11 | 4940 | ||
| NM_173163.2 | NP_775186.1 | 11 | 4925 | |||
| NM_173165.2 | NP_775188.1 | 10 | 4836 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NFATC3| Gene Symbol: NFATC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004555.3 | NP_004546.1 | 11 | 4940 | ||
| NM_173163.2 | NP_775186.1 | 11 | 4925 | |||
| NM_173165.2 | NP_775188.1 | 10 | 4836 | |||