siRNA Details
| Gene Symbol: ESRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BU934976.1 | - | 1 | 209 | ||
| Gene Symbol: NFATC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004555.3 | NP_004546.1 | 11 | 5803 | ||
| NM_173163.2 | NP_775186.1 | 11 | 5788 | |||
| NM_173165.2 | NP_775188.1 | 10 | 5699 | |||

ESRP2
Genomic Map