siRNA Details
| Gene Symbol: PHEX-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046639.1 | 1 | 956 | |||
| GenBank mRNA | AK095185.1 | - | 1 | 956 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PHEX-AS1| Gene Symbol: PHEX-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046639.1 | 1 | 956 | |||
| GenBank mRNA | AK095185.1 | - | 1 | 956 | ||