siRNA Details
| Gene Symbol: ZNF691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017001401.1 | XP_016856890.1 | 1 | 1501 | ||
| XM_017001403.1 | XP_016856892.1 | 1 | 1500 | |||
| GenBank mRNA | AK095184.1 | - | 1 | 1559 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF691| Gene Symbol: ZNF691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017001401.1 | XP_016856890.1 | 1 | 1501 | ||
| XM_017001403.1 | XP_016856892.1 | 1 | 1500 | |||
| GenBank mRNA | AK095184.1 | - | 1 | 1559 | ||