siRNA Details
| Gene Symbol: SIRT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI969140.1 | - | 1 | 79 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC064478.1 | - | 1 | 671 | Chr. 6 - 13614111 - 13615155 [-] on Build GRCh38 | |

SIRT5
Genomic Map