siRNA Details
| Gene Symbol: LOC101929259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120424.1 | 3 | 1203 | |||
| GenBank mRNA | AK093174.1 | - | 3 | 1201 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929259| Gene Symbol: LOC101929259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120424.1 | 3 | 1203 | |||
| GenBank mRNA | AK093174.1 | - | 3 | 1201 | ||