siRNA Details
| Gene Symbol: LOC101929259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120424.1 | 2, 3 | 241 | |||
| GenBank mRNA | AK093174.1 | - | 2, 3 | 241 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB025349.1 | - | 2, 3 | 241 | Chr. 13 - 45106279 - 45112002 [+] on Build GRCh38 | |

LOC101929259
Genomic Map