siRNA Details
| Gene Symbol: LOC100130899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039988.1 | 1 | 4123 | |||
| GenBank mRNA | AK126297.1 | - | 1 | 3975 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | C00661.1 | - | 2 | 187 | Chr. 4 - 119135849 - 119158047 [+] on Build GRCh38 | |

LOC100130899
Genomic Map