siRNA Details
| Gene Symbol: LOC100130899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039988.1 | 1 | 1226 | |||
| GenBank mRNA | AK126297.1 | - | 1 | 1078 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100130899| Gene Symbol: LOC100130899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039988.1 | 1 | 1226 | |||
| GenBank mRNA | AK126297.1 | - | 1 | 1078 | ||