siRNA Details
Gene Symbol: LOC100130899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039988.1 | 1 | 3124 | |||
GenBank mRNA | AK126297.1 | - | 1 | 2976 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100130899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039988.1 | 1 | 3124 | |||
GenBank mRNA | AK126297.1 | - | 1 | 2976 |