siRNA Details
| Gene Symbol: OLMALINC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK023036.1 | - | 2 | 1895 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CV569873.1 | - | 1 | 317 | Chr. 10 - 100382624 - 100383249 [+] on Build GRCh38 | |

OLMALINC
Genomic Map