siRNA Details
| Gene Symbol: LOC100128437 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126077.1 | - | 1 | 934 | ||
| Gene Symbol: LOC105369723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135029.1 | 2 | 434 | |||
| GenBank mRNA | BI771000.1 | - | 2 | 434 | ||

LOC100128437
Genomic Map