siRNA Details
| Gene Symbol: CYP4F12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_117085.1 | 10 | 1591 | |||
| GenBank mRNA | AK074412.1 | - | 5 | 2875 | ||
| AK075435.1 | - | 10 | 1519 | |||
| AL832171.1 | - | 10 | 2480 | |||
| DR422336.1 | - | 1 | 26 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK091995.1 | - | 2025 | Chr. - - [-] on Build | ||

CYP4F12
Genomic Map