siRNA Details
| Gene Symbol: AHSA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011532541.2 | XP_011530843.1 | 8 | 2431 | ||
| XM_011532543.2 | XP_011530845.1 | 9 | 2715 | |||
| XM_017003378.1 | XP_016858867.1 | 9 | 2390 | |||
| GenBank mRNA | AK091998.1 | - | 1 | 1607 | ||

AHSA2
Genomic Map