siRNA Details
| Gene Symbol: SECISBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127655.1 | - | 1 | 2072 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB282642.1 | - | 1 | 208 | Chr. 9 - 89354986 - 89355536 [+] on Build GRCh38 | |

SECISBP2
Genomic Map