siRNA Details
| Gene Symbol: LOC100506258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103855.1 | 3 | 333 | |||
| GenBank mRNA | AK128569.1 | - | 5 | 1571 | ||
| BC040358.1 | - | 3 | 309 | |||
| DA959881.1 | - | 3 | 317 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG177687.1 | - | 3 | 321 | Chr. 11 - 7427711 - 7465822 [-] on Build GRCh38 | |

LOC100506258
Genomic Map